Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | JAZF1 antisense RNA 1 | JAZF1-AS1 | -483.2kb | Sense | Chr. 7 - 28180457 - 28241377 on NCBI Build GRCh38 | |
| Downstream | TLR4 interactor with leucine rich repeats | TRIL | 227.7kb | Anti-sense | Chr. 7 - 28953358 - 28958413 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map