Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | UBE2E1 antisense RNA 1 | UBE2E1-AS1 | -49.4kb | Sense | Chr. 3 - 23804024 - 23806905 on NCBI Build GRCh38 | |
| Downstream | NFKB inhibitor interacting Ras like 1 | NKIRAS1 |
KBRAS1, kappaB-Ras1 |
35.1kb | Sense | Chr. 3 - 23889536 - 23917046 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map