Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | Wilms tumor 1 associated protein | WTAP | Mum2 | Sense | Chr. 6 - 159726680 - 159756319 on NCBI Build GRCh38 | |
| Downstream | uncharacterized LOC100129518 | LOC100129518 | 29.1kb | Anti-sense | Chr. 6 - 159760258 - 159762332 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map