| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02001096.hg.1 |
ENST00000295092.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02001096.hg.1 |
NM_145175.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02001096.hg.1 |
TC02000087.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200006779.hg.1 |
ENST00000295092.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200006779.hg.1 |
ENST00000295092.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200006779.hg.1 |
ENST00000295092.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200006779.hg.1 |
ENST00000295092.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200006779.hg.1 |
ENST00000295092.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200006779.hg.1 |
ENST00000295092.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200006779.hg.1 |
ENST00000295092.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200006779.hg.1 |
ENST00000295092.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200006779.hg.1 |
ENST00000295092.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200006779.hg.1 |
ENST00000295092.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200006779.hg.1 |
ENST00000295092.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200006779.hg.1 |
ENST00000295092.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200006779.hg.1 |
ENST00000331243.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200006779.hg.1 |
ENST00000331243.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200006779.hg.1 |
ENST00000331243.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200006779.hg.1 |
ENST00000331243.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200006779.hg.1 |
ENST00000331243.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200006779.hg.1 |
ENST00000331243.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200006779.hg.1 |
ENST00000331243.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200006779.hg.1 |
ENST00000331243.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200006779.hg.1 |
ENST00000331243.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200006779.hg.1 |
ENST00000331243.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200006779.hg.1 |
ENST00000331243.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200006779.hg.1 |
ENST00000331243.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200006779.hg.1 |
ENST00000464947.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200006779.hg.1 |
ENST00000464947.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200006779.hg.1 |
ENST00000464947.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200006779.hg.1 |
ENST00000464947.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200006779.hg.1 |
ENST00000464947.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200006779.hg.1 |
ENST00000464947.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200006779.hg.1 |
ENST00000497769.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200006779.hg.1 |
ENST00000497769.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200006779.hg.1 |
ENST00000497769.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200006779.hg.1 |
ENST00000497769.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200006779.hg.1 |
ENST00000497769.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200006779.hg.1 |
ENST00000497769.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200006779.hg.1 |
NM_001291410.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200006779.hg.1 |
NM_001291410.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200006779.hg.1 |
NM_001291410.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200006779.hg.1 |
NM_001291410.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200006779.hg.1 |
NM_001291410.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200006779.hg.1 |
NM_001291410.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200006779.hg.1 |
NM_145175.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200006779.hg.1 |
NM_145175.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200006779.hg.1 |
NM_145175.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200006779.hg.1 |
NM_145175.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200006779.hg.1 |
NM_145175.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200006779.hg.1 |
NM_145175.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200006779.hg.1 |
NM_145175.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200006779.hg.1 |
NM_145175.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200006779.hg.1 |
NM_145175.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200006779.hg.1 |
NM_145175.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200006779.hg.1 |
NM_145175.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200006779.hg.1 |
NM_145175.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200146453.hg.1 |
ENST00000295092.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200146453.hg.1 |
NM_145175.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200146453.hg.1 |
ENST00000295092.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200146453.hg.1 |
NM_145175.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200146453.hg.1 |
TC0200006779.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0200006779.hg.1 |
ENST00000295092.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200006779.hg.1 |
ENST00000295092.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200006779.hg.1 |
ENST00000295092.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200006779.hg.1 |
ENST00000295092.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200006779.hg.1 |
ENST00000295092.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200006779.hg.1 |
ENST00000295092.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200006779.hg.1 |
ENST00000295092.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200006779.hg.1 |
ENST00000295092.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200006779.hg.1 |
ENST00000295092.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200006779.hg.1 |
ENST00000295092.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200006779.hg.1 |
ENST00000295092.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200006779.hg.1 |
ENST00000295092.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200006779.hg.1 |
ENST00000331243.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200006779.hg.1 |
ENST00000331243.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200006779.hg.1 |
ENST00000331243.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200006779.hg.1 |
ENST00000331243.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200006779.hg.1 |
ENST00000331243.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200006779.hg.1 |
ENST00000331243.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200006779.hg.1 |
ENST00000331243.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200006779.hg.1 |
ENST00000331243.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200006779.hg.1 |
ENST00000331243.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200006779.hg.1 |
ENST00000331243.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200006779.hg.1 |
ENST00000331243.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200006779.hg.1 |
ENST00000331243.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200006779.hg.1 |
ENST00000464947.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200006779.hg.1 |
ENST00000464947.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200006779.hg.1 |
ENST00000464947.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200006779.hg.1 |
ENST00000464947.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200006779.hg.1 |
ENST00000464947.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200006779.hg.1 |
ENST00000464947.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200006779.hg.1 |
ENST00000497769.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200006779.hg.1 |
ENST00000497769.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200006779.hg.1 |
ENST00000497769.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200006779.hg.1 |
ENST00000497769.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200006779.hg.1 |
ENST00000497769.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200006779.hg.1 |
ENST00000497769.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200006779.hg.1 |
NM_001291410.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200006779.hg.1 |
NM_001291410.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200006779.hg.1 |
NM_001291410.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200006779.hg.1 |
NM_001291410.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200006779.hg.1 |
NM_001291410.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200006779.hg.1 |
NM_001291410.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200006779.hg.1 |
NM_145175.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200006779.hg.1 |
NM_145175.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200006779.hg.1 |
NM_145175.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200006779.hg.1 |
NM_145175.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200006779.hg.1 |
NM_145175.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200006779.hg.1 |
NM_145175.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200006779.hg.1 |
NM_145175.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200006779.hg.1 |
NM_145175.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200006779.hg.1 |
NM_145175.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200006779.hg.1 |
NM_145175.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200006779.hg.1 |
NM_145175.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200006779.hg.1 |
NM_145175.3 |
Refseq |