Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | DnaJ heat shock protein family (Hsp40) member C5 beta | DNAJC5B | CSP-beta | -69.8kb | Sense | Chr. 8 - 66014983 - 66100520 on NCBI Build GRCh38 |
| Downstream | corticotropin releasing hormone | CRH |
CRF, CRH1 |
4.0kb | Anti-sense | Chr. 8 - 66176377 - 66186685 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map