| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017594.hg.1 |
ENST00000366713.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017594.hg.1 |
ENST00000366713.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017594.hg.1 |
ENST00000496552.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017594.hg.1 |
ENST00000496552.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017594.hg.1 |
NR_103539.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017594.hg.1 |
NR_103539.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017594.hg.1 |
NR_103540.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017594.hg.1 |
NR_103540.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100017594.hg.1 |
ENST00000366713.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100017594.hg.1 |
ENST00000366713.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100017594.hg.1 |
ENST00000496552.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100017594.hg.1 |
ENST00000496552.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100017594.hg.1 |
NR_103539.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100017594.hg.1 |
NR_103539.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100017594.hg.1 |
NR_103540.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100017594.hg.1 |
NR_103540.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100233035.hg.1 |
ENST00000366713.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100233035.hg.1 |
NR_103540.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100233035.hg.1 |
ENST00000366713.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100233035.hg.1 |
NR_103540.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100233035.hg.1 |
TC0100017594.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0100233033.hg.1 |
ENST00000366713.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100233033.hg.1 |
ENST00000496552.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100233033.hg.1 |
NR_103539.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100233033.hg.1 |
NR_103540.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100233033.hg.1 |
ENST00000366713.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100233033.hg.1 |
ENST00000496552.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100233033.hg.1 |
NR_103539.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100233033.hg.1 |
NR_103540.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100233033.hg.1 |
TC0100017594.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0100233034.hg.1 |
ENST00000496552.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100233034.hg.1 |
NR_103539.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100233034.hg.1 |
ENST00000496552.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100233034.hg.1 |
NR_103539.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100233034.hg.1 |
TC0100017594.hg.1 |
Clariom D TC |
| Human Genome U133 Plus 2.0 Array |
1569300_PM_at |
- |
- |
| Human Genome U133 Plus 2.0 Array |
1569300_at |
- |
- |
| Human Genome U133 Plus 2.0 Array |
238188_at |
- |
- |
| Human Genome U133 Plus 2.0 Array |
238188_PM_at |
- |
- |