Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | LOC400927-CSNK1E readthrough | LOC400927-CSNK1E | Sense | Chr. 22 - 38290691 - 38398522 on NCBI Build GRCh38 | ||
| Downstream | TPTE and PTEN homologous inositol lipid phosphatase pseudogene | LOC400927 | 9.3kb | Sense | Chr. 22 - 38344665 - 38398926 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map