Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | long intergenic non-protein coding RNA 942 | LINC00942 | -52.2kb | Anti-sense | Chr. 12 - 1500491 - 1504424 on NCBI Build GRCh38 | |
| Downstream | F-box and leucine rich repeat protein 14 | FBXL14 | Fbl14 | 7.3kb | Sense | Chr. 12 - 1565993 - 1594189 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map