Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | GPR75-ASB3 readthrough | GPR75-ASB3 | -19.0kb | Sense | Chr. 2 - 53670293 - 53860033 on NCBI Build GRCh38 | |
| Downstream | acylphosphatase 2 | ACYP2 |
ACYM, ACYP |
233.7kb | Anti-sense | Chr. 2 - 53970838 - 54305300 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map