Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | PDS5 cohesin associated factor A | PDS5A |
PIG54, SCC-112, SCC112 |
Sense | Chr. 4 - 39822863 - 39977956 on NCBI Build GRCh38 | |
| Downstream | acyl-CoA thioesterase 7 pseudogene | LOC344967 | 88.5kb | Sense | Chr. 4 - 40042917 - 40057199 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map