Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
HPS6, biogenesis of lysosomal organelles complex 2 subunit 3 |
HPS6 | BLOC2S3 | -19.7kb | Anti-sense | Chr. 10 - 102065367 - 102068038 on NCBI Build GRCh38 |
| Downstream | LIM domain binding 1 | LDB1 |
CLIM-2, CLIM2, LDB-1, NLI |
16.5kb | Sense | Chr. 10 - 102102100 - 102120473 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map