Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | RIB43A domain with coiled-coils 2 | RIBC2 |
C22orf11, TRIB |
-135.2kb | Sense | Chr. 22 - 45413691 - 45432422 on NCBI Build GRCh38 |
| Downstream | long intergenic non-protein coding RNA 1589 | LINC01589 | TCONS_00029353 | 20.6kb | Anti-sense | Chr. 22 - 45604432 - 45605647 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map