Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
small nucleolar RNA, H/ACA box 11B |
SNORA11B | -20.1kb | Sense | Chr. 14 - 91126426 - 91126552 on NCBI Build GRCh38 | |
| Downstream | G protein-coupled receptor 68 | GPR68 |
GPR12A, OGR1 |
84.4kb | Anti-sense | Chr. 14 - 91232532 - 91264581 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map