Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | ribonuclease H1 | RNASEH1 |
H1RNA, PEOB2, RNH1 |
Sense | Chr. 2 - 3531813 - 3558371 on NCBI Build GRCh38 | |
| Downstream | RNASEH1 antisense RNA 1 | RNASEH1-AS1 | 13.8kb | Anti-sense | Chr. 2 - 3558386 - 3561750 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map