Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | transmembrane protein 45B | TMEM45B | -214.0 | Sense | Chr. 11 - 129815781 - 129860003 on NCBI Build GRCh38 | |
| Downstream | nuclear factor related to kappaB binding protein | NFRKB | INO80G | 2.9kb | Anti-sense | Chr. 11 - 129863773 - 129895596 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map