Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | coiled-coil domain containing 57 | CCDC57 | -17.5kb | Anti-sense | Chr. 17 - 82101458 - 82212892 on NCBI Build GRCh38 | |
| Downstream | microRNA 6787 | MIR6787 | hsa-mir-6787 | 5.3kb | Sense | Chr. 17 - 82236668 - 82236728 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map