Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | interleukin 1 beta | IL1B |
IL-1, IL1-BETA, IL1F2 |
-3.5kb | Anti-sense | Chr. 2 - 112829758 - 112836842 on NCBI Build GRCh38 |
| Downstream | interleukin 37 | IL37 |
FIL1, FIL1(ZETA), FIL1Z, IL-1F7, IL-1H, IL-1H4, IL-1RP1, IL-37, IL1F7, IL1H4, IL1RP1 |
66.9kb | Sense | Chr. 2 - 112908889 - 112918882 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map