Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | nuclear apoptosis inducing factor 1 | NAIF1 |
C9orf90, bA379C10.2 |
-9.8kb | Anti-sense | Chr. 9 - 128061233 - 128067321 on NCBI Build GRCh38 |
| Downstream | SLC25A25 antisense RNA 1 | SLC25A25-AS1 | 31.6kb | Anti-sense | Chr. 9 - 128111171 - 128118734 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map