Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | Yip1 domain family member 4 | YIPF4 |
FinGER4, Nbla11189 |
-188.5kb | Sense | Chr. 2 - 32277889 - 32307929 on NCBI Build GRCh38 |
| Downstream | microRNA 558 | MIR558 |
MIRN558, hsa-mir-558 |
33.4kb | Sense | Chr. 2 - 32532153 - 32532246 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map