Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | HSPE1-MOB4 readthrough | HSPE1-MOB4 | HSPE1-PHOCN | Sense | Chr. 2 - 197499997 - 197553699 on NCBI Build GRCh38 | |
| Downstream |
MOB family member 4, phocein |
MOB4 |
2C4D, CGI-95, MOB1, MOB3, MOBKL3, PHOCN, PREI3 |
14.0kb | Sense | Chr. 2 - 197515571 - 197553699 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map