Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | GA binding protein transcription factor alpha subunit | GABPA |
E4TF1-60, E4TF1A, NFT2, NRF2, NRF2A, RCH04A07 |
-270.8kb | Sense | Chr. 21 - 25734947 - 25772460 on NCBI Build GRCh38 |
| Downstream | cysteine and tyrosine rich 1 antisense RNA 1 | CYYR1-AS1 | 362.1kb | Sense | Chr. 21 - 26393635 - 26569252 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map