Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | uncharacterized LOC105379183 | LOC105379183 | -8.0kb | Anti-sense | Chr. 5 - 133968689 - 133970223 on NCBI Build GRCh38 | |
| Downstream | uncharacterized protein C7orf73-like | LOC101060024 | 101.3kb | Sense | Chr. 5 - 134081411 - 134082833 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map