Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | FUS RNA binding protein | FUS |
ALS6, ETM4, FUS1, HNRNPP2, POMP75, TLS |
-1.3kb | Sense | Chr. 16 - 31179444 - 31194871 on NCBI Build GRCh38 |
| Downstream | PYD and CARD domain containing | PYCARD |
ASC, CARD5, TMS, TMS-1, TMS1 |
4.5kb | Anti-sense | Chr. 16 - 31201486 - 31202776 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map