Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | mitochondrial ribosomal protein L42 | MRPL42 |
HSPC204, L31MT, L42MT, MRP-L31, MRP-L42, MRP-S32, MRPL31, MRPS32, PTD007, RPML31, S32MT |
-38.7kb | Anti-sense | Chr. 12 - 93467490 - 93503772 on NCBI Build GRCh38 |
| Downstream | SOCS2 antisense RNA 1 | SOCS2-AS1 | 19.7kb | Sense | Chr. 12 - 93565628 - 93571398 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map