Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | zinc finger FYVE-type containing 26 | ZFYVE26 |
FYVE-CENT, SPG15 |
-281.5kb | Anti-sense | Chr. 14 - 67728892 - 67817293 on NCBI Build GRCh38 |
| Downstream | uncharacterized LOC100996664 | LOC100996664 | 505.6kb | Anti-sense | Chr. 14 - 68625527 - 68628787 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map