Assay Details
| Gene Symbol: MAP3K8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017015709.1 | XP_016871198.1 | 1 | 1392 | 80 | |
| GenBank mRNA | BC022398.1 | - | 1 | 51 | 80 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | golgin A2 pseudogene 6 | GOLGA2P6 | -63.5kb | Anti-sense | Chr. 10 - 30364327 - 30371907 on NCBI Build GRCh38 | |
| Downstream | lysozyme like 2 | LYZL2 | LYZD2 | 169.6kb | Anti-sense | Chr. 10 - 30606222 - 30629761 on NCBI Build GRCh38 |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_017015709.1 transcripts.
Set Membership: |
|

MAP3K8
Genomic Map