Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | small nucleolar RNA host gene 9 | SNHG9 | NCRNA00062 | Sense | Chr. 16 - 1964996 - 1965504 on NCBI Build GRCh38 | |
| Downstream | ring finger protein 151 | RNF151 | 1.3kb | Sense | Chr. 16 - 1966639 - 1968975 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map