Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | microRNA 8064 | MIR8064 | hsa-mir-8064 | -33.9kb | Anti-sense | Chr. 3 - 52846463 - 52846552 on NCBI Build GRCh38 |
| Downstream | Scm-like with four mbt domains 1 | SFMBT1 |
RU1, SFMBT, hSFMBT |
18.8kb | Anti-sense | Chr. 3 - 52899209 - 53046655 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map