Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | RNF219 antisense RNA 1 | RNF219-AS1 | POU4F1-AS1 | -14.1kb | Anti-sense | Chr. 13 - 78054855 - 78617325 on NCBI Build GRCh38 |
| Downstream | long intergenic non-protein coding RNA 331 | LINC00331 | NCRNA00331 | 154.4kb | Sense | Chr. 13 - 78787319 - 78840050 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map