Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | uncharacterized LOC100996291 | LOC100996291 | -5.3kb | Sense | Chr. 17 - 78261349 - 78278492 on NCBI Build GRCh38 | |
| Downstream | suppressor of cytokine signaling 3 | SOCS3 |
ATOD4, CIS3, Cish3, SOCS-3, SSI-3, SSI3 |
72.3kb | Sense | Chr. 17 - 78356777 - 78360079 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map