Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | retinal degeneration 3 | RD3 |
C1orf36, LCA12 |
-47.6kb | Sense | Chr. 1 - 211476522 - 211492917 on NCBI Build GRCh38 |
| Downstream | solute carrier family 30 member 1 | SLC30A1 |
ZNT1, ZRC1 |
33.5kb | Sense | Chr. 1 - 211575039 - 211578757 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map