Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | uncharacterized LOC105377777 | LOC105377777 | -135.8kb | Anti-sense | Chr. 8 - 878741 - 882930 on NCBI Build GRCh38 | |
| Downstream | uncharacterized LOC286083 | LOC286083 | 276.3kb | Anti-sense | Chr. 8 - 1296034 - 1302607 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map