Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | ankyrin repeat and sterile alpha motif domain containing 1B | ANKS1B |
AIDA, AIDA-1, ANKS2, EB-1, EB1, cajalin-2 |
-47.6kb | Sense | Chr. 12 - 98729904 - 99984727 on NCBI Build GRCh38 |
| Downstream | UHRF1 binding protein 1 like | UHRF1BP1L | SHIP164 | 3.1kb | Sense | Chr. 12 - 100026251 - 100142864 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map