Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | homeobox C8 | HOXC8 |
HOX3, HOX3A |
-7.1kb | Sense | Chr. 12 - 54009106 - 54012763 on NCBI Build GRCh38 |
| Downstream | microRNA 615 | MIR615 |
MIRN615, hsa-mir-615, mir-615 |
11.4kb | Sense | Chr. 12 - 54033950 - 54034045 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map