Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | U2AF homology motif (UHM) kinase 1 | UHMK1 |
KIS, KIST, P-CIP2 |
-30.6kb | Anti-sense | Chr. 1 - 162497174 - 162529629 on NCBI Build GRCh38 |
| Downstream | UDP-N-acetylglucosamine pyrophosphorylase 1 | UAP1 |
AGX, AGX1, AGX2, SPAG2 |
193.0 | Anti-sense | Chr. 1 - 162561440 - 162599843 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map