Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | lemur tyrosine kinase 2 | LMTK2 |
AATYK2, BREK, KPI-2, KPI2, LMR2, PPP1R100, cprk, hBREK |
Anti-sense | Chr. 7 - 98106868 - 98209633 on NCBI Build GRCh38 | |
| Downstream | basic helix-loop-helix family member a15 | BHLHA15 |
BHLHB8, MIST1 |
85.8kb | Anti-sense | Chr. 7 - 98212254 - 98212959 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map