Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | microRNA 4423 | MIR4423 | mir-4423 | -18.6kb | Anti-sense | Chr. 1 - 85133794 - 85133873 on NCBI Build GRCh38 |
| Downstream | synapse defective Rho GTPase homolog 2 | SYDE2 | 3.5kb | Sense | Chr. 1 - 85157673 - 85201725 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map