Assay Details
| Gene Symbol: COL4A2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC068210.1 | - | 1 | 451 | 62 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | COL4A2 antisense RNA 1 | COL4A2-AS1 | -3.2kb | Anti-sense | Chr. 13 - 110502576 - 110508179 on NCBI Build GRCh38 | |
| Downstream |
RAB20, member RAS oncogene family |
RAB20 | 10.0kb | Anti-sense | Chr. 13 - 110523066 - 110561737 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

COL4A2
Genomic Map