Assay Details
| Gene Symbol: ZNF800 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005250181.3 | XP_005250238.1 | 6 | 13697 | 97 | |
| GenBank mRNA | BX537723.1 | - | 1 | 3851 | 97 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | glutamate metabotropic receptor 8 | GRM8 |
GLUR8, GPRC1H, MGLUR8, mGlu8 |
-106.5kb | Sense | Chr. 7 - 126438598 - 127253294 on NCBI Build GRCh38 |
| Downstream | uncharacterized LOC100506682 | LOC100506682 | 112.1kb | Anti-sense | Chr. 7 - 127476883 - 127485804 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

ZNF800
Genomic Map