Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | prune homolog 2 | PRUNE2 |
BMCC1, BNIPXL, C9orf65, KIAA0367 |
Anti-sense | Chr. 9 - 76611376 - 76906220 on NCBI Build GRCh38 | |
| Downstream | prostate cancer associated 3 (non-protein coding) | PCA3 |
DD3, NCRNA00019, PCAT3 |
79.7kb | Sense | Chr. 9 - 76764436 - 76787569 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map