Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | NFIA antisense RNA 1 | NFIA-AS1 | -98.2kb | Anti-sense | Chr. 1 - 61248945 - 61253518 on NCBI Build GRCh38 | |
| Downstream | sepiapterin reductase (7,8-dihydrobiopterin:NADP+ oxidoreductase) pseudogene | MGC34796 | 297.9kb | Sense | Chr. 1 - 61654242 - 61656128 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map