Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | DDRGK domain containing 1 | DDRGK1 |
C20orf116, UFBP1, dJ1187M17.3 |
Sense | Chr. 20 - 3190350 - 3204697 on NCBI Build GRCh38 | |
| Downstream | inosine triphosphatase | ITPA |
C20orf37, HLC14-06-P, ITPase, My049, NTPase, dJ794I6.3 |
14.8kb | Anti-sense | Chr. 20 - 3208868 - 3227449 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map