Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | tubulin alpha like 3 | TUBAL3 | -37.4kb | Sense | Chr. 10 - 5393098 - 5404830 on NCBI Build GRCh38 | |
| Downstream | uncharacterized LOC105376379 | LOC105376379 | 40.3kb | Sense | Chr. 10 - 5483489 - 5493864 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map