Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | uncharacterized LOC284933 | LOC284933 | -39.1kb | Anti-sense | Chr. 22 - 48538900 - 48547387 on NCBI Build GRCh38 | |
| Downstream | microRNA 4535 | MIR4535 | 191.9kb | Sense | Chr. 22 - 48780295 - 48780353 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map