Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | myosin heavy chain gene cluster antisense RNA | MYHAS | -33.0kb | Anti-sense | Chr. 17 - 10383132 - 10625540 on NCBI Build GRCh38 | |
| Downstream | SCO1 cytochrome c oxidase assembly protein | SCO1 | SCOD1 | 21.3kb | Sense | Chr. 17 - 10679770 - 10697568 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map