Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | miR-17-92a-1 cluster host gene | MIR17HG |
C13orf25, FGLDS2, LINC00048, MIHG1, MIRH1, MIRHG1, NCRNA00048, miR-17-92 |
-976.9kb | Anti-sense | Chr. 13 - 91347820 - 91354575 on NCBI Build GRCh38 |
| Downstream | GPC5 antisense RNA 2 | GPC5-AS2 | 7.9kb | Sense | Chr. 13 - 92339794 - 92352207 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map