Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | RNF219 antisense RNA 1 | RNF219-AS1 | POU4F1-AS1 | Anti-sense | Chr. 13 - 78054855 - 78617325 on NCBI Build GRCh38 | |
| Downstream | POU class 4 homeobox 1 | POU4F1 |
BRN3A, Oct-T1, RDC-1, brn-3A |
174.9kb | Sense | Chr. 13 - 78599092 - 78603560 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map