Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | Yip1 domain family member 4 | YIPF4 |
FinGER4, Nbla11189 |
Anti-sense | Chr. 2 - 32277889 - 32307929 on NCBI Build GRCh38 | |
| Downstream | baculoviral IAP repeat containing 6 | BIRC6 |
APOLLON, BRUCE |
78.9kb | Anti-sense | Chr. 2 - 32356907 - 32618899 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map