Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | betaine--homocysteine S-methyltransferase 2 | BHMT2 | -27.4kb | Sense | Chr. 5 - 79069724 - 79090074 on NCBI Build GRCh38 | |
| Downstream | long intergenic non-protein coding RNA 485 | LINC00485 | 328.7kb | Sense | Chr. 12 - 102809283 - 102824399 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map