Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | microRNA 6132 | MIR6132 | hsa-mir-6132 | -18.7kb | Sense | Chr. 7 - 117020211 - 117020319 on NCBI Build GRCh38 |
| Downstream | ST7 antisense RNA 2 | ST7-AS2 |
ST7AS2, ST7OT2 |
32.7kb | Anti-sense | Chr. 7 - 117072072 - 117145592 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map